missing translation for 'onlineSavingsMsg'
Learn More

Glycine methyl ester hydrochloride, 99%

Catalog No. AAA140830E
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA140830E 2500 g
AAA1408322 100 g
AAA1408336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA140830E Supplier Thermo Scientific Chemicals Supplier No. A140830E
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5680-79-5 | C3H8ClNO2 | 125.55 g/mol

Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

Solubility
Soluble in water (2% w/v) and methanol (50 mg/ml).

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.

Chemical Identifiers

CAS 5680-79-5
Molecular Formula C3H8ClNO2
Molecular Weight (g/mol) 125.55
MDL Number MFCD00012870
InChI Key COQRGFWWJBEXRC-UHFFFAOYSA-N
Synonym glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
PubChem CID 122755
IUPAC Name methyl 2-aminoacetate;hydrochloride
SMILES [H+].[Cl-].COC(=O)CN

Specifications

Melting Point ∼175°C (decomposition)
Beilstein 3593644
Solubility Information Soluble in water (2% w/v) and methanol (50mg/ml).
Density 1
Quantity 2500 g
Formula Weight 125.56
Percent Purity 99%
Chemical Name or Material Glycine methyl ester hydrochloride
EINECSNumber 227-139-1
TSCA No

RUO – Research Use Only