missing translation for 'onlineSavingsMsg'
Learn More

Glycine methyl ester hydrochloride, 99%

CAS: 5680-79-5 | C3H8ClNO2 | 125.55 g/mol

Supplier:  Thermo Scientific Chemicals A1408322

 View more versions of this product

Catalog No. AAA1408322


Only null left

Description

Description

Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glycine methyl ester hydrochloride a non-essential amino acid used for human development.

Solubility
Soluble in water (2% w/v) and methanol (50 mg/ml).

Notes
Hygroscopic. Store in cool, dry place in tightly closed containers. Store away from oxidizing agent and water/moisture.
Specifications

Specifications

∼175°C (decomposition)
5680-79-5
MFCD00012870
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
COQRGFWWJBEXRC-UHFFFAOYSA-N
1
100 g
122755
99%
Glycine methyl ester hydrochloride
C3H8ClNO2
3593644
Soluble in water (2% w/v) and methanol (50mg/ml).
[H+].[Cl-].COC(=O)CN
methyl 2-aminoacetate;hydrochloride
125.55
125.56

Chemical Identifiers

5680-79-5
125.55
COQRGFWWJBEXRC-UHFFFAOYSA-N
122755
[H+].[Cl-].COC(=O)CN
C3H8ClNO2
MFCD00012870
glycine methyl ester hydrochloride, h-gly-ome.hcl, methyl 2-aminoacetate hydrochloride, methyl glycinate hydrochloride, glycine, methyl ester, hydrochloride, methyl glycinate hcl, methyl aminoacetate hydrochloride, glycine methyl ester hcl, glycine methyl ester hydrochloride salt, aminoacetic acid methyl ester hydrochloride
methyl 2-aminoacetate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 227-139-1

TSCA : No

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only