missing translation for 'onlineSavingsMsg'
Learn More

Glycine Crystalline MP Biomedicals

Supplier:  MP Biomedicals Inc 0210057001

 View more versions of this product

Catalog No. ICN10057001


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Glycine
C2H5NO2
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
NCC(O)=O
2-aminoacetic acid
75.07
CHEBI:15428
Crystalline
56-40-6
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
1.16 g/mL
1 kg
750
75.07

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions