missing translation for 'onlineSavingsMsg'
Learn More

Glycine Crystalline MP Biomedicals

$142.18 - $1985.75

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10057001
View Documents
MP Biomedicals Inc
0210057001
1 kg
Each for $142.18
Only null left
 
ICN10057005
View Documents
MP Biomedicals Inc
0210057005
5 kg
Each for $536.36
Only null left
 
ICN10057025
View Documents
MP Biomedicals Inc
0210057025
25 kg
Each for $1,985.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O

Specifications

56-40-6
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
1.16 g/mL
1 kg
750
75.07
Glycine
C2H5NO2
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
NCC(O)=O
2-aminoacetic acid
75.07
CHEBI:15428
Crystalline
Videos
SDS
Documents

Documents