missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, Crystalline, MP Biomedicals
$73.32 - $165.63
Chemical Identifiers
| CAS | 56-40-6 |
|---|---|
| Molecular Formula | C2H5NO2 |
| Molecular Weight (g/mol) | 75.07 |
| MDL Number | MFCD00008131 |
| InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
| Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
| PubChem CID | 750 |
| ChEBI | CHEBI:15428 |
| IUPAC Name | 2-aminoacetic acid |
| SMILES | NCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN1946811
|
MP Biomedicals Inc
021946811 |
100 g |
N/A
|
|
|||||
|
ICN19468101
|
MP Biomedicals Inc
0219468101 |
1 kg |
N/A
|
|
|||||
Chemical Identifiers
| 56-40-6 | |
| 75.07 | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| 750 | |
| 2-aminoacetic acid |
| C2H5NO2 | |
| MFCD00008131 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| CHEBI:15428 | |
| NCC(O)=O |
Specifications
| 56-40-6 | |
| MFCD00008131 | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| 2-aminoacetic acid | |
| 75.07 | |
| CHEBI:15428 | |
| Cell Culture | |
| Glycine, Free Acid |
| C2H5NO2 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| NCC(O)=O | |
| 100 g | |
| 750 | |
| 75.07 | |
| Crystals |
Safety and Handling
Recommended Storage : Room temperature