missing translation for 'onlineSavingsMsg'
Learn More

Glycine, Crystalline, MP Biomedicals

Supplier:  MP Biomedicals Inc 0219468101

 View more versions of this product

Catalog No. ICN19468101

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Glycine
56-40-6
MFCD00008131
DHMQDGOQFOQNFH-UHFFFAOYSA-N
2-aminoacetic acid
75.07
CHEBI:15428
Cell Culture
Free Acid
C2H5NO2
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
NCC(O)=O
1 kg
750
75.07
Crystals

Chemical Identifiers

56-40-6
75.07
DHMQDGOQFOQNFH-UHFFFAOYSA-N
750
2-aminoacetic acid
C2H5NO2
MFCD00008131
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
CHEBI:15428
NCC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room temperature

SDS Documents

Documents

Promotions

Promotions