missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, ACS, 98.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor GL15812KG
Specifications
| Glycine | |
| 56-40-6 | |
| 5e-05 | |
| Poly Bottle | |
| 5e-05 | |
| 98.5% |
| 5e-05 | |
| 1 | |
| 2e-05 | |
| 0.001 | |
| 12 kg | |
| ACS |
Chemical Identifiers
| 56-40-6 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.