missing translation for 'onlineSavingsMsg'
Learn More

Glycine, ACS, 98.5%, Spectrum™ Chemical

Catalog No. p-7165626
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610741 100 g Poly Bottle
18610745 500 g Poly Bottle
18610740 1 kg Poly Bottle
18610743 2.5 kg Poly Bottle
18610742 12 kg Poly Bottle
18610744 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610741 Supplier Spectrum Chemical Mfg Cor Supplier No. GL158100GM
Only null left

Chemical Identifiers

CAS 56-40-6

Specifications

Ammonium 5e-05
CAS Max % 1
Chloride 5e-05
Heavy Metals 2e-05
Packaging Poly Bottle
Residue after Ignition 0.001
Sulfate 5e-05
Quantity 100 g
Percent Purity 98.5%
Grade ACS
Chemical Name or Material Glycine
Show More Show Less

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.