missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycine, 10% (w/v), Ricca Chemical
Supplier: Ricca Chemical Company 328330
Specifications
| Glycine | |
| 89.0,8.91 | |
| C2H5NO2 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| NCC(O)=O | |
| 75.07 | |
| CHEBI:15428 | |
| Aqueous Solution | |
| 10% (w/v) |
| 56-40-6,7732-18-5 | |
| 93.0,9.28 | |
| MFCD00008131 | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| 2-aminoacetic acid | |
| 750 | |
| Poly Drum | |
| 30 gal. |
Chemical Identifiers
| 56-40-6 | |
| 75.07 | |
| DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
| 750 | |
| 2-aminoacetic acid |
| C2H5NO2 | |
| MFCD00008131 | |
| glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
| CHEBI:15428 | |
| NCC(O)=O |