missing translation for 'onlineSavingsMsg'
Learn More

Glycinamide Hydrochloride 98.0+%, TCI America™

$48.13 - $300.33

Chemical Identifiers

CAS 1668-10-6
Molecular Formula C2H7ClN2O
Molecular Weight (g/mol) 110.541
MDL Number MFCD00013008
InChI Key WKNMKGVLOWGGOU-UHFFFAOYSA-N
Synonym glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
PubChem CID 2723639
IUPAC Name 2-aminoacetamide;hydrochloride
SMILES C(C(=O)N)N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
G025325G
View Documents
TCI America
G025325G
25 g
Each for $48.13
Only null left
 
G0253100G
missing translation for 'viewDocuments'
TCI America
G0253100G
100 g
Each for $105.89
Only null left
 
G0253500G
missing translation for 'viewDocuments'
TCI America
G0253500G
500 g
Each for $300.33
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1668-10-6
110.541
WKNMKGVLOWGGOU-UHFFFAOYSA-N
2723639
C(C(=O)N)N.Cl
C2H7ClN2O
MFCD00013008
glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
2-aminoacetamide;hydrochloride

Specifications

1668-10-6
C2H7ClN2O
25 g
WKNMKGVLOWGGOU-UHFFFAOYSA-N
2-aminoacetamide;hydrochloride
2723639
≥98.0% (T)
Glycinamide Hydrochloride
White-Yellow
MFCD00013008
glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride
C(C(=O)N)N.Cl
110.541
110.54
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents