accessibility menu, dialog, popup

UserName

HClH2NNH2O

CAS RN 1668-10-6

IUPAC Name: hydrogen 2-aminoacetamide chloride
Synonyms: hydrogen glycinamid chloride
Molecular Weight (g/mol): 110.54
Molecular Formula: C2H7ClN2O
InChi Key: WKNMKGVLOWGGOU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC(N)=O
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (5)
  • (5)
Quantity 
  • (4)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (8)
  • (5)
  • (5)
  • (4)
  • (4)
  • (1)
  • (1)
  • (7)