missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycinamide Hydrochloride 98.0+%, TCI America™
Supplier: TCI America G025325G
Specifications
| Glycinamide Hydrochloride | |
| White-Yellow | |
| MFCD00013008 | |
| glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride | |
| C(C(=O)N)N.Cl | |
| 110.541 | |
| 110.54 | |
| Crystalline Powder |
| 1668-10-6 | |
| C2H7ClN2O | |
| 25 g | |
| WKNMKGVLOWGGOU-UHFFFAOYSA-N | |
| 2-aminoacetamide;hydrochloride | |
| 2723639 | |
| ≥98.0% (T) |
Chemical Identifiers
| 1668-10-6 | |
| 110.541 | |
| WKNMKGVLOWGGOU-UHFFFAOYSA-N | |
| 2723639 | |
| C(C(=O)N)N.Cl |
| C2H7ClN2O | |
| MFCD00013008 | |
| glycinamide hydrochloride, 2-aminoacetamide hydrochloride, glycine amide hydrochloride, h-gly-nh2.hcl, glycinamidehydrochloride, acetamide, 2-amino-, monohydrochloride, glycinamide hcl, glycineamide hcl, glycineamide hydrochloride, glycinamid hydrochloride | |
| 2-aminoacetamide;hydrochloride |
Safety and Handling
TSCA : No