missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ultrapure, HPLC Grade

Catalog No. AA38988M1
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38988M1 4 x 1 L
AA38988K2 1 L
AA38988K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA38988M1 Supplier Thermo Scientific Chemicals Supplier No. 038988M1

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Glycerol is used as emulsifier, thickening agent in cosmetics and foods, plasticizer in packaging, paints and paper industry, preservative in food and beverages. It is used in pharmaceutical preparations to improve smoothness and providing lubrication. It is also used in epichlorohydrin and mono propylene glycol production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.26
Boiling Point 182°C (20 mm)
Flash Point 160°C (320°F)
Odor Odorless
Refractive Index 1.474
Quantity 4 x 1 L
Beilstein 635685
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 92.09
Grade HPLC
Physical Form Viscous Liquid
Chemical Name or Material Glycerol, HPLC Grade
Show More Show Less
Hazard Category H319: Causes serious eye irritation.
EINECSNumber 200-289-5
RTECSNumber MA8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only