missing translation for 'onlineSavingsMsg'
Learn More

Glycerol (Ultra Pure 99.5%)

Supplier:  MP Biomedicals Inc 04800688

 View more versions of this product

Catalog No. ICN800688


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Glycerol (Ultra Pure 99.5%)
C3H8O3
1 L
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
Ultrapure
56-81-5
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
OCC(O)CO
92.09
CHEBI:17754

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions