missing translation for 'onlineSavingsMsg'
Learn More

Glycerol Solution, puriss p.a., 86 to 89% (T), Solstice

Catalog No. 60048451
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60048451 1 L Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60048451 Supplier Honeywell Inc Supplier No. 497821L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Puriss. p.a., 86-89% (T)

  • Solstice has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 20°C
Density 1.252g/mL at 25°C
Boiling Point 290°C
Flash Point 160°C (320°F)
Packaging Poly bottle
Refractive Index 1.454
Linear Formula HOCH2CH(OH)CH2OH · aq
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Formula Weight 92.09g/mol (anhydrous basis)
Percent Purity 86 to 89% (T)
Grade ACS
Physical Form Liquid
Chemical Name or Material Glycerol solution
Show More Show Less
RTECSNumber 200-289-5