missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Solution, puriss., Meets analytical specification of Ph. Eur. BP, 84 to 88%, Honeywell™ Riedel-de Haen™
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Specifications
| Color | Colorless |
| Density | 1.252g/mL at 25°C |
| Boiling Point | 290°C |
| Flash Point | 160°C (320°F) |
| Ignition Residue | ≤0.01% (as SO4) |
| Packaging | Poly Bottle |
| Refractive Index | n20/D 1.449-1.455 |
| Linear Formula | HOCH2CH(OH)CH2OH · aq |
| Quantity | 5 L |
| UN Number | NONH for all modes of transport |
| Show More |
Safety and Handling
| EINECSNumber | 200-289-5 |
| RTECSNumber | MA8050000 |