missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol Solution, puriss., Meets analytical specification of Ph. Eur. BP, 84 to 88%, Honeywell™ Riedel-de Haen™
Puriss., meets analytical specification of BP, European Pharmacopoeia, 84-88%
Supplier: Honeywell Chemicals 155244X5L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
| Glycerol Solution | |
| 56-81-5 | |
| 1.252g/mL at 25°C | |
| 160°C (320°F) | |
| Poly Bottle | |
| n20/D 1.449-1.455 | |
| MFCD00004722 | |
| NONH for all modes of transport | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| OCC(O)CO | |
| 92.09 | |
| CHEBI:17754 | |
| 84 to 88% | |
| Liquid |
| Meets analytical specification of Ph. Eur. BP | |
| Colorless | |
| 290°C | |
| ≤0.01% (as SO4) | |
| C3H8O3 | |
| HOCH2CH(OH)CH2OH · aq | |
| 4 x 5 L | |
| 635685 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 753 | |
| 92.09g/mol (anhydrous basis) | |
| puriss. |
Chemical Identifiers
| 56-81-5 | |
| 92.09 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| 753 | |
| propane-1,2,3-triol |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Safety and Handling
EINECSNumber : 200-289-5
RTECSNumber : MA8050000