missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, puriss., Meets analytical specification of Ph. Eur. BP, USP, FCC., Honeywell™ Riedel-de Haen™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E422, anhydrous, 99.0-101.0% (alkalimetric)
$325.87 - $11,238.68
Chemical Identifiers
| CAS | 56-81-5 |
|---|---|
| Molecular Formula | C3H8O3 |
| Molecular Weight (g/mol) | 92.09 |
| MDL Number | MFCD00004722 |
| InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
| Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
| PubChem CID | 753 |
| ChEBI | CHEBI:17754 |
| IUPAC Name | propane-1,2,3-triol |
| SMILES | OCC(O)CO |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
60044885
|
Honeywell-Riedel-de Haen
155231L |
1 L | Poly Bottle |
Each for $325.87
|
|
||||
|
60044888
|
Honeywell-Riedel-de Haen
155235L |
5 L | Poly Bottle |
Each for $1,274.68
|
|
||||
|
60044889
|
Honeywell-Riedel-de Haen
155236X1L |
6 x 1 L | Poly Bottle |
Case of 6 Each for $1,646.66
|
|
||||
|
60044887
|
Honeywell-Riedel-de Haen
155234X5L |
4 x 5 L | Poly Bottle |
Case of 4 Each for $4,164.95
|
|
||||
|
60044886
|
Honeywell-Riedel-de Haen
15523250KG |
250 kg | Poly Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Specifications
| 56-81-5 | |
| Colorless | |
| 290°C | |
| ≤0.01% (as SO4) | |
| C3H8O3 | |
| HOCH2CH(OH)CH2OH | |
| NONH for all modes of transport | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| OCC(O)CO | |
| 92.09 | |
| 3.1 (vs air) | |
| 1mmHg (20°C) | |
| 99.0 to 101.0% (calc. on H2O free substance) | |
| Liquid |
| 18°C | |
| 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C | |
| 160°C (320°F) | |
| 698°F | |
| n20/D 1.474(lit.) | |
| MFCD00004722 | |
| 635685 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 753 | |
| CHEBI:17754 | |
| 92.09g/mol | |
| puriss. | |
| Glycerol, Meets analytical specification of Ph. Eur. BP, USP, FCC. |
Safety and Handling
EINECSNumber : 200-289-5
RTECSNumber : MA8050000