missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glycerol, OmniPur™, Calbiochem™,

Catalog No. 47501L
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47501L 1 L
47604L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 47501L Supplier MilliporeSigma™ Supplier No. 47501L

For Molecular biology

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 19.8°C
Color Colorless
Density 1.260-1.263g/mL at 25°C
Boiling Point 290°C
Flash Point 159.85°C (319.7°F)
Ignition Point 369.85°C (697.7°F)
Odor Odorless
Quantity 1 L
Solubility Information Partially soluble in water
Formula Weight 92.1
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material Glycerol
Grade Molecular biology grade
Show More Show Less
Recommended Storage Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.