missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, Molecular Biology Grade

Catalog No. AAJ61059K2
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61059K2 1 L
AAJ61059AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61059K2 Supplier Thermo Scientific Chemicals Supplier No. J61059K2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Useful as a cryoprotectant. Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component. | CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Useful as a cryoprotectant.Glycerol is evolved in the storage and concentration of enzymes. It plays a vital role in sample preparation and gel formation for polyacrylamide gel electrophoresis. It is also used to aid in casting gradient gels, protein stabilizer and storage buffer component. Further, it is used as a solvent, emollient, pharmaceutical agent and sweetening agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Density 1.26
Boiling Point 182°C (20 mmHg)
Flash Point 160°C (320°F)
Packaging Plastic bottle
Refractive Index 1.474
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
Beilstein 635685
Sensitivity Hygroscopic
Specific Gravity 1.26
Solubility Information Miscible with water and alcohol. Partially miscible with ether and ethyl acetate. Immiscible with chloroform.
Formula Weight 92.09
Grade Molecular Biology
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
EINECSNumber 200-289-5
RTECSNumber MA8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only