missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Glycerol, Molecular Biology Grade, Calbiochem™,

Catalog No. 356352100ML
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
356352100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 356352100ML Supplier MilliporeSigma™ Supplier No. 356352100ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

High-purity glycerol suitable for molecular biology applications including electrophoresis.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Quantity 100 mL
Formula Weight 92.1
Percent Purity ≥99%
Grade Molecular Biology
Physical Form Liquid
Chemical Name or Material Glycerol
RTECSNumber MA8050000
Recommended Storage 15° to 30°C (59° to 86°F)