missing translation for 'onlineSavingsMsg'
Learn More

Glycerol (Molecular Biology), Fisher BioReagents™

Catalog No. BP2291
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2291 1 L Glass Bottle
BP2294 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2291 Supplier Fisher BioReagents Supplier No. FLBP229 1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For low-temperature storage of enzymes and bacterial cultures.

For low-temperature storage of enzymes and bacterial cultures

Glycerol is used for low-temperature storage of enzymes and bacterial cultures, purification of proteins, and as a component in electrophoresis loading buffers.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Decomposition Information >290°C
Melting Point 18°C
Density 1.261g/cm³
pH 5
Boiling Point 290°C
Assay Percent Range ≥99.5 %
Packaging Glass Bottle
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
Merck Index 15, 4520
Vapor Pressure 0.003mbar at 50°C
Formula Weight 92.09
Percent Purity ≥99.5%
Grade DNase-, RNase- and Protease-Free
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:1
Flammability:1
Instability:0
Recommended Storage RT