missing translation for 'onlineSavingsMsg'
Learn More

Glycerol (Glycerin), Purified, Reagents

Catalog No. 01337231
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337231 1 L
01337230 20 L
01337229 4 L
01337228 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337231 Supplier Labchem Inc Supplier No. C22105001C
Only null left

Glycerol (Glycerin), Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym 1,2,3-Propanetriol, Glycerine
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18.2°C
Color Colorless
Boiling Point 544°C
Melting Range 18.2°C
Packaging Glass Bottle
Quantity 1 L
Specific Gravity 1.2613
Grade Pure
Physical Form Liquid
Chemical Name or Material Glycerol (Glycerin), Purified