missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol (Glycerin), ACS Reagent, Reagents
Specifications
Specifications
| Melting Point | 18.2°C |
| Color | Colorless |
| Boiling Point | 544°C |
| Packaging | Poly Drum |
| Quantity | 200 L |
| Specific Gravity | 1.2613 |
| Grade | ACS |
| Physical Form | Liquid |
| Chemical Name or Material | Glycerol (Glycerin), ACS Reagent |