Learn More
Glycerol (Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific BPG33200
Specifications
| Glycerol | |
| >290°C | |
| 1.261g/cm³ | |
| 290°C | |
| ≥99.5 % | |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| OCC(O)CO | |
| 92.09 | |
| CHEBI:17754 | |
| 0.003mbar at 50°C | |
| Certified ACS |
| 56-81-5 | |
| 18°C | |
| 5 | |
| 0.005% max. | |
| Poly Drum | |
| HOCH2CH(OH)CH2OH | |
| 200 L | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 753 | |
| 92.09 | |
| ≥99.5% | |
| Viscous Liquid |
Chemical Identifiers
| 56-81-5 | |
| 92.09 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| 753 | |
| propane-1,2,3-triol |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.
NFPA
Health:1
Flammability:1
Instability:0