missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, Anhydrous, Ultrapure Bioreagent, J.T. Baker™
Description
- For enzyme stabilization, freezing and electrophoresis buffers
Specifications
Specifications
| Flash Point | 199.00°C |
| Packaging | HDPE Pressure Bottom Poly Bottle |
| Quantity | 500 mL |
| Specific Gravity | 1L = 1.26kg |
| Formula Weight | 92.1 |
| Grade | Not Available |
| Physical Form | Liquid |
| Chemical Name or Material | Glycerol, Anhydrous |