missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ACS, 99.5+%

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

$205.44 - $529.25

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA36646K2
View Documents
Thermo Scientific Chemicals
036646K2
1 L N/A
Only null left
 
AA36646K7
View Documents
Thermo Scientific Chemicals
036646K7
4 L N/A
Only null left
 
Description

Description

Glycerol is widely used in cosmetics, especially for production of soaps and lotions, solvents and emulsifiers. It may also be found in inks, adhesives and pharmaceuticals. It plays an important role in the manufacture of plastics and resins. Glycerol bonded with silica gel plays an important role for effective removal of traces of Co(II) from aqueous solutions. Also useful for optical clearing of gastric tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO

Specifications

56-81-5
1.26 g/cm3
160°C (320°F)
>99.5%
1.474
1 L
Hygroscopic
Miscible in water,methanol,ethanol and 2-propanol. Immiscible in ether,benzene and chloroform.
OCC(O)CO
92.09
CHEBI:17754
<0.1 hPa (0mm Hg) at 20°C
Viscous Liquid
18°C
182°C (20 mm)
400°C (752°F)
C3H8O3
MFCD00004722
635685
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
ACS
Glycerol
Videos
Safety and Handling

Safety and Handling

H319: Causes serious eye irritation.

EINECSNumber : 200-289-5

RTECSNumber : MA8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only