missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ACS, 99.5+%

Catalog No. AA36646K2
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36646K2 1 L
AA36646K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36646K2 Supplier Thermo Scientific Chemicals Supplier No. 036646K2

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Glycerol is widely used in cosmetics, especially for production of soaps and lotions, solvents and emulsifiers. It may also be found in inks, adhesives and pharmaceuticals. It plays an important role in the manufacture of plastics and resins. Glycerol bonded with silica gel plays an important role for effective removal of traces of Co(II) from aqueous solutions. Also useful for optical clearing of gastric tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Density 1.26 g/cm3
Boiling Point 182°C (20 mm)
Flash Point 160°C (320°F)
Ignition Point 400°C (752°F)
Assay Percent Range >99.5%
Refractive Index 1.474
Quantity 1 L
Beilstein 635685
Sensitivity Hygroscopic
Solubility Information Miscible in water,methanol,ethanol and 2-propanol. Immiscible in ether,benzene and chloroform.
Vapor Pressure <0.1 hPa (0mm Hg) at 20°C
Formula Weight 92.09
Grade ACS
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less
Hazard Category H319: Causes serious eye irritation.
EINECSNumber 200-289-5
RTECSNumber MA8050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only