missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99+%

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

Supplier:  Thermo Scientific Chemicals A162050F

 View more versions of this product

Catalog No. AAA162050F


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Glycerol is used as a solvent, sweetener, preservative, antifreezing agent, thickening agent in liqueurs and humectants in the food industry. It is also used in the preparation of pharmaceutical and personal care products, thereby improving the smoothness and provides lubrication. It is an active component in glycerin soap. It finds application in cough syrups, elixirs and expectorants, toothpaste, soaps, mouthwashes, skin care products, shaving cream, hair care products, and water-based personal lubricants. It is used as a starting material for the preparation of nitroglycerin, allyl iodidie, epichlorohydrin and glycerine acetate. In medical field, it is also used as a laxative and a temporary reduction in the internal pressure of the eye. In addition to this, it is involved to aid in casting gradient gels, protein stabilizer and storage buffer component.

Solubility
Miscible with water and alcohol. Partially miscible with ether and ethyl acetate. Immiscible with chloroform.

Notes
Hygroscopic. Incompatible with perchloric acid, lead oxide, acetic anhydride, nitrobenzene, chlorine, peroxides, strong acids and strong bases.
Specifications

Specifications

Glycerol
18°C
182°C (20 mmHg)
Odorless
1.474
MFCD00004722
635685
14,4484
Miscible with water and alcohol. Partially miscible with ether and ethyl acetate. Immiscible with chloroform.
OCC(O)CO
92.09
CHEBI:17754
≥99%
56-81-5
1.26
160°C (320°F)
C3H8O3
HOCH2CH(OH)CH2OH
2500 mL
Hygroscopic
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

Warning

EINECSNumber : 200-289-5

RTECSNumber : MA8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only