missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99% max., Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19399680
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19399680 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19399680 Supplier MP Biomedicals Inc Supplier No. 0219399680
Only null left

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Density 1.2613g/cm3 at 20°C
Quantity 100 mL
Formula Weight 92.09
Percent Purity 99% max.
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material Glycerol