accessibility menu, dialog, popup
UserName
Fisher Scientific Logo
EN
chevron-down
Select Language
English
Français
Search
Order Status
Quick Order
Support
Offers
chevron-down
Featured
Spotlight
Offers
Flex Dollars
New Lab Start-up
View All
Sign In
chevron-down
chevron-down
missing translation for 'onlineSavingsMsg'
Learn More
Home
Products
Biochemical Reagents
Carbohydrates
Sugar alcohols
ICN19468080
Structure Search
Print
Glycerol, ≥99%, Cell Culture Reagent, MP Biomedicals™
Catalog No.
ICN19468080
Change view
Chip view icon
List view icon
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
Quantity
ICN19468080
100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No.
ICN19468080
Supplier
MP Biomedicals Inc
Supplier No.
0219468080
Sign In
or
Register
to check your price and availability.
Quantity
Only null left
Add to Cart
Add to Cart
Description
Specifications
Product Suggestions
Videos
Documents
Certificates
Safety and Handling
Safety Services
Promotions
Specifications
Chemical Identifiers
CAS
56-81-5
Molecular Formula
C3H8O3
Molecular Weight (g/mol)
92.09
MDL Number
MFCD00004722
InChI Key
PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID
753
ChEBI
CHEBI:17754
IUPAC Name
propane-1,2,3-triol
SMILES
OCC(O)CO
Specifications
Quantity
100 mL
Product Suggestions
Documents