missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99.91%, MP Biomedicals™

$86.19 - $123.03

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN15119490
View Documents
MP Biomedicals Inc
0215119490
500 mL
Each for $86.19
Only null left
 
ICN15119491
View Documents
MP Biomedicals Inc
0215119491
1 L
Each for $123.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO

Specifications

56-81-5
1.2613g/cm3 at 20°C
C3H8O3
500 mL
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
Glycerol
Colorless
0.9991
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
OCC(O)CO
92.09
CHEBI:17754
Viscous Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'