missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, 99.91%, MP Biomedicals™
Specifications
Specifications
| Color | Colorless |
| Density | 1.2613g/cm3 at 20°C |
| Assay Percent Range | 0.9991 |
| Quantity | 500 mL |
| Formula Weight | 92.09 |
| Physical Form | Viscous Liquid |
| Chemical Name or Material | Glycerol |