missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, >99%, Highly Purified, MP Biomedicals™

Catalog No. ICN19520480
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19520480 100 mL
ICN19468090 500 mL
ICN19520491 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19520480 Supplier MP Biomedicals Inc Supplier No. 0219520480

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Density 1.2613 g/cu cm at 20°C (Lit.)
Odor Odorless
Assay Percent Range >99%
Quantity 100 mL
Formula Weight 92.09
Grade Highly Purified
Physical Form Syrupy Liquid
Chemical Name or Material Glycerol
Recommended Storage Store at Room Temperature (15 to 30°C).