missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, 99.6%, ACS reagent

Catalog No. AC410980100
Change view
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410980100 10 L Plastic drum
AC410980025 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC410980100 Supplier Thermo Scientific Chemicals Supplier No. 410980100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Decomposition Information >290°C
Melting Point 18.0°C
Density 1.2610g/mL
Boiling Point 290.0°C
Flash Point 160°C
Ignition Residue 0.005% max.
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.4716
Linear Formula HOCH2CH(OH)CH2OH
Quantity 10 L
Beilstein 01, 502
Specific Gravity 1.261
Merck Index 15, 4520
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Viscosity 1069 mPa.s (20°C)
Vapor Pressure 0.003mbar at 50°C
Formula Weight 92.09
Percent Purity 99.6%
Grade ACS Reagent
Physical Form Liquid
Chemical Name or Material Glycerol
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-289-5
RTECSNumber MA8050000
TSCA TSCA

RUO – Research Use Only