missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ≥99.5%, Molecular biology reagent grade, MP Biomedicals™

Catalog No. ICN19399690
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19399690 500 mL
ICN19399691 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19399690 Supplier MP Biomedicals Inc Supplier No. 0219399690
Only null left

Glycerol is a simple polyol compound which is a viscous liquid that is widely used in pharmaceutical formulations. It has three hydroxyl groups that are responsible for its solubility in water and its hygroscopic nature

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Density 1.2613g/cm3
Assay Percent Range ≥99.5%
Quantity 500 mL
Formula Weight 92.09
Grade Molecular Biology Reagent
Physical Form Liquid
Chemical Name or Material Glycerol
EINECSNumber 200-289-5