missing translation for 'onlineSavingsMsg'
Learn More

Glycerol, ≥99.5%, Molecular biology reagent grade, MP Biomedicals™

Glycerol is a simple polyol compound which is a viscous liquid that is widely used in pharmaceutical formulations. It has three hydroxyl groups that are responsible for its solubility in water and its hygroscopic nature

Supplier:  MP Biomedicals Inc 0219399691

 View more versions of this product

Catalog No. ICN19399691

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Glycerol
1.2613g/cm3
C3H8O3
1 L
PEDCQBHIVMGVHV-UHFFFAOYSA-N
propane-1,2,3-triol
753
92.09
Liquid
56-81-5
≥99.5%
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
OCC(O)CO
92.09
CHEBI:17754
Molecular Biology Reagent

Chemical Identifiers

56-81-5
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
753
propane-1,2,3-triol
C3H8O3
MFCD00004722
glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
CHEBI:17754
OCC(O)CO
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 200-289-5

SDS
Documents

Documents

Promotions

Promotions