missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glycerol, ACS Reagent, ≥99.5%, Honeywell Riedel-de Haën™
ACS Reagent,≥99.5%
$974.13 - $974.13
Chemical Identifiers
| CAS | 56-81-5 |
|---|---|
| Molecular Formula | C3H8O3 |
| Molecular Weight (g/mol) | 92.09 |
| MDL Number | MFCD00004722 |
| InChI Key | PEDCQBHIVMGVHV-UHFFFAOYSA-N |
| Synonym | glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn |
| PubChem CID | 753 |
| ChEBI | CHEBI:17754 |
| IUPAC Name | propane-1,2,3-triol |
| SMILES | OCC(O)CO |
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Chemical Identifiers
| 56-81-5 | |
| 92.09 | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| 753 | |
| propane-1,2,3-triol |
| C3H8O3 | |
| MFCD00004722 | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| CHEBI:17754 | |
| OCC(O)CO |
Specifications
| 56-81-5 | |
| Colorless | |
| Neutral | |
| 160°C (320°F) | |
| Odorless | |
| C3H8O3 | |
| MFCD00004722 | |
| NONH for all modes of transport | |
| glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn | |
| PEDCQBHIVMGVHV-UHFFFAOYSA-N | |
| propane-1,2,3-triol | |
| 3.1 (vs air) | |
| CHEBI:17754 | |
| 1mmHg (20°C) | |
| Liquid |
| 18°C | |
| 1.25g/mL | |
| 290°C (1013 hPa) | |
| 698°F | |
| Poly bottle | |
| HOCH2CH(OH)CH2OH | |
| 4 L | |
| 635685 | |
| Completely miscible in water | |
| OCC(O)CO | |
| 92.09 | |
| 753 | |
| 92.09g/mol | |
| ≥99.5% | |
| Glycerol |
Safety and Handling
RTECSNumber : 200-289-5