missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, Reagent Grade, LabChem™

Catalog No. LC148502
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC148502 1 L Poly Bottle
LC148501 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC148502 Supplier LabChem Supplier No. LC148502
Only null left

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Max % 100
Decomposition Information fume; Carbon monoxide; Carbon dioxide
Melting Point 20°C
Color Colorless
Density 1.2613g/mL
Boiling Point 182°C
Flash Point 160°C
Packaging Poly Bottle
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
Specific Gravity 1.2613g/mL
Solubility Information Soluble in water
Formula Weight 92.09
Grade Reagent
Physical Form Liquid
Chemical Name or Material Glycerin
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
EINECSNumber 200-289-5
Recommended Storage Room Temperature