missing translation for 'onlineSavingsMsg'
Learn More

Glycerin, ACS Reagent Grade, Ricca Chemical

Catalog No. RSOG00201A
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RSOG00201A 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOG00201A Supplier Ricca Chemical Company Supplier No. RSOG00201A
Only null left

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless
Boiling Point 544°C
Packaging Natural Poly Bottle
Quantity 1 L
Percent Purity Min. 99.5%
Grade ACS
Physical Form Liquid
Chemical Name or Material Glycerin
ShelfLife 60 months