missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Glutathione Sepharose™ 4 Fast Flow Media
Description
- Glutathione coupled to Sepharose 4 Fast Flow via 10-carbon spacer arm
- Protein binding capacity: ≈10mg recombinant GST/mL media (flowrate dependent)
- Binding capacity and high flow properties allow scale-up
- Mild elution conditions preserve protein antigenicity and function
- Stable in all commonly used aqueous buffers
- pH stability: 3 to 12
Specifications
Specifications
| Binding Capacity | >10 mg/mL |
| Description | Glutathione Sepharose 4 Fast Flow is an affinity resin especially suited for purification of GST-tagged proteins in a batch mode or when scale up is needed. |
| Flow Rate | <450 cm/hr. |
| Kit Contents | 10 Carbon Spacer Arm, 4% Agarose, 90 μm Particle Size |
| Matrix | Highly cross-linked 4% agarose |
| pH Range | 3 to 12 |
| Support Type | 4% Highly Cross-linked Agarose |
| Content And Storage | 4°C to 30°C, 20% Ethanol |
| Max. Operating Pressure | base matrix 150-250 cm/h, 100 kPa, XK 50/60 column, bed height 25 cm |
| Particle Size | 90 μm |
| Show More |