missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Glutathione Sepharose™ 4 Fast Flow Media

Catalog No. 45000286
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45000286 100 mL
45000285 25 mL
45000287 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 45000286 Supplier Cytiva Supplier No. 17513202

Single-step purification of GST-tagged proteins and other glutathione S-transferase or glutathione-dependent proteins. Glutathione Sepharose 4 Fast Flow Media is used for the purification of GST-tagged proteins from bacterial lysates and other S-transferases or glutathione-dependent proteins.

  • Glutathione coupled to Sepharose 4 Fast Flow via 10-carbon spacer arm
  • Protein binding capacity: ≈10mg recombinant GST/mL media (flowrate dependent)
  • Binding capacity and high flow properties allow scale-up
  • Mild elution conditions preserve protein antigenicity and function
  • Stable in all commonly used aqueous buffers
  • pH stability: 3 to 12

Specifications

Binding Capacity >10 mg/mL
Description Glutathione Sepharose 4 Fast Flow is an affinity resin especially suited for purification of GST-tagged proteins in a batch mode or when scale up is needed.
Flow Rate <450 cm/hr.
Kit Contents 10 Carbon Spacer Arm, 4% Agarose, 90 μm Particle Size
Matrix Highly cross-linked 4% agarose
pH Range 3 to 12
Support Type 4% Highly Cross-linked Agarose
Content And Storage 4°C to 30°C, 20% Ethanol
Max. Operating Pressure base matrix 150-250 cm/h, 100 kPa, XK 50/60 column, bed height 25 cm
Particle Size 90 μm
Quantity 100 mL
Type Chromatography Media
Ligand Type Glutathione and 10-carbon linker arm
Show More Show Less