missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaric dialdehyde, 8% solution in water
CAS: 111-30-8 | C5H8O2 | 100.12 g/mol
Supplier: Thermo Scientific Chemicals 410971000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Solution | |
| 111-30-8 , 7732-18-5 | |
| Glass bottle | |
| 100 g | |
| OHC(CH2)3CHO | |
| 01, 776 | |
| 15, 4508 | |
| SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
| pentanedial | |
| 3485 | |
| 100.12 |
| Glutaric dialdehyde | |
| 1.0160g/mL | |
| 0.918 to 1.123 (20°/20°C) | |
| C5H8O2 | |
| MFCD00007025 | |
| 01,411 | |
| glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
| O=CCCCC=O | |
| 100.12 | |
| CHEBI:64276 | |
| 7 to 9 vol.% |
RUO – Research Use Only