missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutaraldehyde (ca. 50% in Water, ca. 5.6mol/L), TCI America™
$34.18 - $87.34
Chemical Identifiers
| CAS | 111-30-8 |
|---|---|
| Molecular Formula | C5H8O2 |
| Molecular Weight (g/mol) | 100.12 |
| MDL Number | MFCD00007025 |
| InChI Key | SXRSQZLOMIGNAQ-UHFFFAOYSA-N |
| Synonym | glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde |
| PubChem CID | 3485 |
| ChEBI | CHEBI:64276 |
| IUPAC Name | pentanedial |
| SMILES | O=CCCCC=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G006825ML
|
TCI America
G006825ML |
25 mL |
Each for $34.18
|
|
|||||
|
G0068500ML
|
TCI America
G0068500ML |
500 mL |
Each for $87.34
|
|
|||||
Chemical Identifiers
| 111-30-8 | |
| 100.12 | |
| SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
| 3485 | |
| pentanedial |
| C5H8O2 | |
| MFCD00007025 | |
| glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
| CHEBI:64276 | |
| O=CCCCC=O |
Specifications
| Clear Liquid at 20°C | |
| Yellow | |
| 25 mL | |
| MFCD00007025 | |
| glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde | |
| O=CCCCC=O | |
| 100.12 | |
| CHEBI:64276 | |
| Glutaraldehyde (ca. 50% in Water, ca. 5.6mol/L) |
| 111-30-8 | |
| 3.0 to 6.0 | |
| C5H8O2 | |
| 2927 | |
| SXRSQZLOMIGNAQ-UHFFFAOYSA-N | |
| pentanedial | |
| 3485 | |
| 100.12 |
Safety and Handling
EINECSNumber : (2)-0509
RTECSNumber : MA2450000
TSCA : Yes