missing translation for 'onlineSavingsMsg'
Learn More

Glutaraldehyde (ca. 50% in Water, ca. 5.6mol/L), TCI America™

Supplier:  TCI America G0068500ML

 View more versions of this product

Catalog No. G0068500ML

Only null left

Description

Description

Specifications

Specifications

Clear Liquid at 20°C
111-30-8
3.0 to 6.0
C5H8O2
2927
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
pentanedial
3485
100.12
Glutaraldehyde (ca. 50% in Water, ca. 5.6mol/L)
Yellow
500 mL
MFCD00007025
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
O=CCCCC=O
100.12
CHEBI:64276

Chemical Identifiers

111-30-8
100.12
SXRSQZLOMIGNAQ-UHFFFAOYSA-N
3485
pentanedial
C5H8O2
MFCD00007025
glutaraldehyde, glutaral, glutaric dialdehyde, cidex, glutardialdehyde, 1,5-pentanedial, sonacide, glutarol, aldesan, glutaric aldehyde
CHEBI:64276
O=CCCCC=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0509

RTECSNumber : MA2450000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions