missing translation for 'onlineSavingsMsg'
Learn More

Glucono delta-Lactone, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7157516
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602924 100 g Poly Bottle
18602925 1 kg Poly Bottle
18602926 5 kg Poly Pail
18602927 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18602924 Supplier Spectrum Chemical Mfg Cor Supplier No. GL107100GM
Only null left

GL107, 90-80-2, C6H10O6

Spectrum™ Chemical Glucono delta-Lactone, FCC is used as a sequestrant, a leavening agent, and an acidifier. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 90-80-2
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00006647
InChI Key PHOQVHQSTUBQQK-UHFFFAOYNA-N
IUPAC Name 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
SMILES OCC1OC(=O)C(O)C(O)C1O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 100 g
Percent Purity 99 to 100.5%
Grade FCC