Learn More
Globe Scientific Dual-Position Snap Caps
For aerobic or anaerobic storage of culture samples
Supplier: Globe Scientific 118118
Description
- Low density polyethylene made, ribbed caps
- Provides leak-resistant seal when snapped into tube
Leave the cap in loose position for aerobic work and in sealed position for anaerobic cultures.
Chemical Identifiers
| Snap Cap | |
| LDPE | |
| Natural |
Specifications
| Snap Cap | |
| LDPE | |
| Natural | |
| 17 mm Test Tubes |