missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Dual-Position Snap Caps

Catalog No. p-4654118
Change view
Click to view available options
For Use With (Equipment):
12 mm Test Tubes
17 mm Test Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22171663 12 mm Test Tubes
21200264 17 mm Test Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22171663 Supplier Globe Scientific Supplier No. 118120

For aerobic or anaerobic storage of culture samples

  • Low density polyethylene made, ribbed caps
  • Provides leak-resistant seal when snapped into tube

Leave the cap in loose position for aerobic work and in sealed position for anaerobic cultures.

Specifications

Product Type Snap Cap
Material LDPE
Color Natural
For Use With (Equipment) 12 mm Test Tubes