Learn More
Glimepiride
A sulfonylurea hypoglecemic agent and potent potassium channel blocker | CAS: 93479-97-1 | C24H34N4O5S | 490.62 g/mol
Supplier: Thermo Scientific Chemicals J6203203
Description
Glimepiride induces the PI3 kinase (PI3K) and Akt pathway, along with insulin receptor substrate-1/2 and endothelial nitric oxide synthase. Glimepiride also increases osteoblast proliferation and differentiation, which is thought to be related to its ability to activate the PI3K and Akt pathway. Furthermore, Glimepiride enhances intrinsic peroxisome proliferator-activated receptor γ activity. Glimepiride also increases protein expression of glucose transports 1 and 4, and is a potent KIR channel blocker. Potent Kir6 (KATP) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac Kir6 channels with an IC50 of 6.8 nM.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsGlimepiride induces the PI3 kinase (PI3K) and Akt pathway, along with insulin receptor substrate-1/2 and endothelial nitric oxide synthase. Glimepiride also increases osteoblast proliferation and differentiation, which is thought to be related to its ability to activate the PI3K and Akt pathway. Furthermore, Glimepiride enhances intrinsic peroxisome proliferator-activated receptor γ activity. Glimepiride also increases protein expression of glucose transports 1 and 4, and is a potent KIR channel blocker. Potent Kir6 (KATP) channel blocker and anti-diabetic agent. Inhibits pinacidil-activated cardiac Kir6 channels with an IC50 of 6.8 nM.
Solubility
Soluble in DMSO
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications
| Glimepiride | |
| 212°C to 215°C | |
| C24H34N4O5S | |
| 1 g | |
| glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin | |
| WIGIZIANZCJQQY-UHFFFAOYSA-N | |
| 3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide | |
| 3476 | |
| Powder |
| 93479-97-1 | |
| White | |
| MFCD00878417 | |
| 14,4440 | |
| Soluble in DMSO | |
| CCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O | |
| 490.62 | |
| 490.62 |
Chemical Identifiers
| 93479-97-1 | |
| 490.62 | |
| WIGIZIANZCJQQY-UHFFFAOYSA-N | |
| 3476 | |
| CCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O |
| C24H34N4O5S | |
| MFCD00878417 | |
| glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin | |
| 3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide |
Safety and Handling
RTECSNumber : UX9363950
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only