missing translation for 'onlineSavingsMsg'
Learn More

Glimepiride 98.0+%, TCI America™

Catalog No. p-7130257
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03951G 1 g
G03955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03951G Supplier TCI America Supplier No. G03951G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93479-97-1
Molecular Formula C24H34N4O5S
Molecular Weight (g/mol) 490.62
MDL Number MFCD00878417
InChI Key WIGIZIANZCJQQY-UHFFFAOYSA-N
Synonym glimepiride, amaryl, glimepiridum, glimepirida, amarel, glimepirid, glimepride, endial, roname, glimepiridum latin
PubChem CID 3476
IUPAC Name 3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide
SMILES CCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O

Specifications

Melting Point 204°C
Color White
Quantity 1 g
Formula Weight 490.62
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Glimepiride
RTECSNumber UX9363950
TSCA No